Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025939.6 | NP_001021110.1 | 11 - 12 | 2840 | 91 | |
NM_001047257.6 | NP_001040722.1 | 3 - 4 | 329 | 91 | ||
NM_062788.7 | NP_495189.2 | 4 - 5 | 530 | 91 | ||
GenBank mRNA | AY091591.1 | - | 11 - 12 | 2801 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |