Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025939.6 | NP_001021110.1 | 7 - 8 | 2324 | 67 | |
GenBank mRNA | AY091591.1 | - | 7 - 8 | 2285 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |