Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062870.7 | NP_495271.1 | 11 - 12 | 1865 | 113 | |
GenBank mRNA | AF104017.1 | - | 11 - 12 | 1846 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |