Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313632.4 | NP_001300561.1 | 4 - 5 | 752 | 72 | |
NM_001313633.3 | NP_001300562.1 | 3 - 4 | 577 | 72 | ||
NM_001313634.3 | NP_001300563.1 | 3 - 4 | 577 | 72 | ||
NM_062884.7 | NP_495285.1 | 4 - 5 | 750 | 72 | ||
GenBank mRNA | AF275253.1 | - | 4 - 5 | 757 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |