Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abch-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062919.5 | NP_495320.1 | 9 - 10 | 1365 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |