Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047288.4 | NP_001040753.1 | 6 - 7 | 1075 | 131 | |
NM_001047289.6 | NP_001040754.1 | 2 - 3 | 463 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |