Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ift-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356863.3 | NP_001343748.1 | 7 - 8 | 1676 | 105 | |
NM_062958.6 | NP_495359.2 | 8 - 9 | 1921 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |