Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aff-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063001.6 | NP_495402.3 | 11 - 12 | 1606 | 111 | |
GenBank mRNA | EF205023.1 | - | 11 - 12 | 1606 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |