Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026783.6 | NP_001021954.1 | 4 - 5 | 593 | 65 | |
NM_063025.9 | NP_495426.1 | 4 - 5 | 593 | 65 | ||
GenBank mRNA | M91371.1 | - | 4 - 5 | 629 | 65 | |
S42187.1 | - | 4 - 5 | 629 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |