Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063050.6 | NP_495451.1 | 17 - 18 | 2413 | 101 | |
GenBank mRNA | AF178755.1 | - | 17 - 18 | 2413 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |