Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026891.6 | NP_001022062.1 | 2 - 3 | 206 | 131 | |
NR_151558.1 | 1 - 2 | 165 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |