Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026889.5 | NP_001022060.1 | 3 - 4 | 273 | 68 | |
NM_001313600.3 | NP_001300529.1 | 2 | 161 | 68 | ||
NM_001313601.3 | NP_001300530.1 | 2 | 134 | 68 | ||
NM_001313602.3 | NP_001300531.1 | 1 - 2 | 44 | 68 | ||
NM_001313604.3 | NP_001300533.1 | 2 | 134 | 68 | ||
NM_001313605.3 | NP_001300534.1 | 1 - 2 | 44 | 68 | ||
NM_001313606.3 | NP_001300535.1 | 2 - 3 | 161 | 68 | ||
NR_132585.1 | 2 - 3 | 161 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |