Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063151.4 | NP_495552.2 | 4 - 5 | 440 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |