Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368597.3 | NP_001355411.1 | 6 | 632 | 113 | |
NM_001368598.4 | NP_001355412.1 | 4 - 5 | 544 | 113 | ||
NM_001383889.1 | 4 - 5 | 626 | 113 | |||
NM_063171.6 | NP_495572.3 | 6 - 7 | 722 | 113 | ||
GenBank mRNA | EF471980.1 | - | 4 - 5 | 542 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |