Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027334.3 | NP_001022505.1 | 3 - 4 | 334 | 92 | |
NM_001381498.1 | NP_001367580.1 | 2 - 3 | 282 | 92 | ||
NM_001381499.2 | NP_001367579.1 | 2 - 3 | 379 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |