Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026975.4 | NP_001022146.1 | 10 - 11 | 1073 | 82 | |
NM_001383891.1 | 8 - 9 | 890 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |