Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alfa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063202.8 | NP_495603.1 | 8 - 9 | 1816 | 99 | |
NM_063203.6 | NP_495604.1 | 7 - 8 | 1821 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |