Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lir-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026926.4 | NP_001022097.1 | 4 - 5 | 544 | 126 | |
NM_001026927.3 | NP_001022098.1 | 2 - 3 | 465 | 126 | ||
GenBank mRNA | AJ130959.1 | - | 3 - 4 | 499 | 126 | |
AJ130960.1 | - | 2 - 3 | 451 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |