Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027149.5 | NP_001022320.1 | 4 - 5 | 2210 | 103 | |
NM_001383900.1 | 4 - 5 | 2211 | 103 | |||
NM_063290.3 | NP_495691.1 | 4 - 5 | 2225 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |