Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfxn-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026851.6 | NP_001022022.1 | 2 - 3 | 568 | 69 | |
NM_001322747.3 | NP_001309542.1 | 1 - 2 | 273 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |