Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027145.3 | NP_001022316.1 | 5 - 6 | 999 | 70 | |
NM_001027146.3 | NP_001022317.1 | 5 - 6 | 911 | 70 | ||
GenBank mRNA | AF063244.1 | - | 5 - 6 | 933 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |