Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027147.5 | NP_001022318.1 | 5 - 6 | 913 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |