Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027088.7 | NP_001022259.1 | 5 - 6 | 1495 | 66 | |
NM_001381521.1 | NP_001368728.1 | 4 - 5 | 1356 | 66 | ||
NM_001383902.1 | 5 - 6 | 1456 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |