Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267227.3 | NP_001254156.1 | 2 - 3 | 469 | 71 | |
NM_001381528.1 | NP_001367210.1 | 2 - 3 | 433 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |