Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267230.4 | NP_001254159.1 | 4 - 5 | 334 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |