Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063421.6 | NP_495822.1 | 2 - 3 | 286 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |