Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063424.7 | NP_495825.1 | 5 - 6 | 945 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |