Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063424.7 | NP_495825.1 | 6 - 7 | 1412 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |