Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026848.5 | NP_001022019.1 | 2 - 3 | 283 | 86 | |
NM_001026849.3 | NP_001022020.1 | 2 - 3 | 279 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |