Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rha-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373779.1 | NP_001360473.1 | 11 - 12 | 3299 | 68 | |
NM_063489.6 | NP_495890.2 | 11 - 12 | 3299 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |