Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267257.3 | NP_001254186.1 | 3 - 4 | 205 | 64 | |
NM_063491.6 | NP_495892.1 | 4 - 5 | 305 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |