Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330955.2 | NP_001317778.1 | 4 - 5 | 629 | 114 | |
NM_001381544.1 | NP_001368599.1 | 4 - 5 | 593 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |