Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063556.8 | NP_495957.1 | 4 - 5 | 733 | 105 | |
GenBank mRNA | AY028165.1 | - | 4 - 5 | 720 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |