Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356885.2 | NP_001343566.1 | 3 | 292 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Both primers and probe map within a single exon |