Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063585.9 | NP_495986.3 | 5 - 6 | 1334 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |