Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377837.1 | NP_001364551.1 | 10 - 11 | 2290 | 78 | |
NM_063636.1 | NP_496037.1 | 10 - 11 | 2296 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |