Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063665.7 | NP_496066.2 | 8 - 9 | 1068 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |