Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063700.4 | NP_496101.1 | 3 - 4 | 675 | 94 | |
NM_063701.4 | NP_496102.1 | 4 - 5 | 861 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |