Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267303.3 | NP_001254232.1 | 1 - 2 | 186 | 63 | |
NM_063745.3 | NP_496146.1 | 11 - 12 | 2874 | 63 | ||
NM_063746.5 | NP_496147.1 | 7 - 8 | 2141 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |