Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027117.6 | NP_001022288.1 | 5 - 6 | 630 | 112 | |
NM_001373787.1 | NP_001359539.1 | 4 - 5 | 511 | 112 | ||
NR_002363.2 | 5 - 6 | 676 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |