Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026968.4 | NP_001022139.1 | 4 - 5 | 530 | 71 | |
NM_001026969.4 | NP_001022140.1 | 5 - 6 | 604 | 71 | ||
NM_001267307.4 | NP_001254236.1 | 5 - 6 | 617 | 71 | ||
NM_001306556.3 | NP_001293485.1 | 1 - 2 | 79 | 71 | ||
GenBank mRNA | AF226156.1 | - | 5 - 6 | 627 | 71 | |
AF233652.1 | - | 4 - 5 | 552 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |