Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063794.1 | NP_496195.1 | 1 - 2 | 114 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |