Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ref-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063803.5 | NP_496204.1 | 3 - 4 | 768 | 84 | |
GenBank mRNA | AF358857.1 | - | 3 - 4 | 790 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |