Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pro-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063823.5 | NP_496224.2 | 6 - 7 | 1194 | 140 | |
GenBank mRNA | EU068465.1 | - | 6 - 7 | 1193 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |