Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063867.3 | NP_496268.2 | 7 - 8 | 1538 | 67 | |
NR_138506.1 | 6 - 7 | 1279 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |