Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ggr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063905.4 | NP_496306.2 | 3 - 4 | 622 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |