Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026867.4 | NP_001022038.2 | 18 - 19 | 2538 | 139 | |
NM_001083157.4 | NP_001076626.2 | 12 - 13 | 1931 | 139 | ||
NM_001129127.4 | NP_001122599.1 | 11 - 12 | 1877 | 139 | ||
NM_001129129.4 | NP_001122601.1 | 16 - 17 | 2360 | 139 | ||
NM_001377853.2 | NP_001364655.1 | 19 - 20 | 2591 | 139 | ||
NM_001377854.2 | NP_001364657.1 | 18 - 19 | 2504 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |