Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373792.1 | NP_001359656.1 | 6 - 7 | 1077 | 87 | |
NM_001381607.1 | NP_001367045.1 | 6 - 7 | 1080 | 87 | ||
NM_064041.5 | NP_496442.1 | 5 - 6 | 797 | 87 | ||
NR_138514.1 | 4 - 5 | 1926 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |