Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064052.8 | NP_496453.1 | 2 - 3 | 279 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |