Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064133.5 | NP_496534.1 | 15 - 16 | 5027 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |